Product Details

SNP ID
rs146548258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:14147263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACCAGCGCAAGTGATTGCTGTA[A/C]TAAATTTGCAATTGTAATCATAATT
Phenotype
MIM: 604070
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DGKB PubMed Links

Gene Details

Gene
DGKB
Gene Name
diacylglycerol kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004080.2 4470 UTR 3 NP_004071.1
NM_145695.2 4470 Intron NP_663733.1
XM_005249628.2 4470 Intron XP_005249685.1
XM_005249629.2 4470 Intron XP_005249686.1
XM_005249630.2 4470 Intron XP_005249687.1
XM_005249631.3 4470 Intron XP_005249688.1
XM_011515153.2 4470 Intron XP_011513455.1
XM_011515154.2 4470 Intron XP_011513456.1
XM_011515156.2 4470 Intron XP_011513458.1
XM_011515157.2 4470 Intron XP_011513459.1
XM_011515158.2 4470 Intron XP_011513460.1
XM_017011783.1 4470 Intron XP_016867272.1
XM_017011784.1 4470 Intron XP_016867273.1
XM_017011785.1 4470 Intron XP_016867274.1
XM_017011786.1 4470 Intron XP_016867275.1
XM_017011787.1 4470 Intron XP_016867276.1
XM_017011788.1 4470 Intron XP_016867277.1
XM_017011789.1 4470 Intron XP_016867278.1
XM_017011790.1 4470 Intron XP_016867279.1
XM_017011791.1 4470 Intron XP_016867280.1
XM_017011792.1 4470 Intron XP_016867281.1

View Full Product Details