Product Details

SNP ID
hCV173280176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73331735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGGAGTTTGGCAACTACCTTTTC[C/T]GCCAGAATCGTTTCTATGATGCCAA
Phenotype
MIM: 604839 MIM: 612548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP6 PubMed Links

Gene Details

Gene
FKBP6
Gene Name
FK506 binding protein 6
There are no transcripts associated with this gene.

Gene
TRIM50
Gene Name
tripartite motif containing 50
There are no transcripts associated with this gene.

View Full Product Details