Product Details

SNP ID
rs147238224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:98293587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGAGTTTCACAGTGGCAAAGGG[G/T]TTTTCTCCGCTGCAGGGGATAAGAA
Phenotype
MIM: 611877 MIM: 615628
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BAIAP2L1 PubMed Links

Gene Details

Gene
BAIAP2L1
Gene Name
BAI1 associated protein 2 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018842.4 1733 Missense Mutation AAA,AAC K490N NP_061330.2
Gene
BRI3
Gene Name
brain protein I3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159491.1 1733 Intron NP_001152963.1
NM_015379.4 1733 Intron NP_056194.1
XM_017011931.1 1733 Intron XP_016867420.1
XM_017011932.1 1733 Intron XP_016867421.1
XM_017011933.1 1733 Intron XP_016867422.1
XM_017011934.1 1733 Intron XP_016867423.1
XM_017011935.1 1733 Intron XP_016867424.1
XM_017011936.1 1733 Intron XP_016867425.1

View Full Product Details