Product Details

SNP ID
rs147544126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:886589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGCAAAACAGTGCAGCAGACTT[A/G]TCCATGCTGGTCCTGGAGTCCCTGG
Phenotype
MIM: 612056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GET4 PubMed Links

Gene Details

Gene
GET4
Gene Name
golgi to ER traffic protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015949.2 347 Silent Mutation TTA,TTG L85L NP_057033.2

View Full Product Details