Product Details

SNP ID
rs147611242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:56102963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACACTCCAGAAACTGTTTGATCT[C/T]ATAGAGGCAAGGCTGCTGCTGCTGT
Phenotype
MIM: 616244 MIM: 172470
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHCHD2 PubMed Links

Gene Details

Gene
CHCHD2
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320327.1 Intron NP_001307256.1
NM_016139.3 Intron NP_057223.1
Gene
PHKG1
Gene Name
phosphorylase kinase catalytic subunit gamma 1
There are no transcripts associated with this gene.

View Full Product Details