Product Details

SNP ID
rs148387469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100220560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGTGCCCAGGGCTCTCTGCCCC[A/G]CCGACTCCTGCCCCCATTCTGCGGG
Phenotype
MIM: 617012 MIM: 608489
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GATS PubMed Links

Gene Details

Gene
GATS
Gene Name
GATS, stromal antigen 3 opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178831.6 850 Intron NP_849153.3
XM_017012153.1 850 Intron XP_016867642.1
XM_017012154.1 850 Intron XP_016867643.1
XM_017012155.1 850 Intron XP_016867644.1
XM_017012156.1 850 Intron XP_016867645.1
XM_017012157.1 850 Intron XP_016867646.1
XM_017012158.1 850 Intron XP_016867647.1
XM_017012159.1 850 Intron XP_016867648.1
Gene
PVRIG
Gene Name
poliovirus receptor related immunoglobulin domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024070.3 850 Silent Mutation CCA,CCG P161P NP_076975.2
XM_011516575.2 850 Silent Mutation CCA,CCG P161P XP_011514877.1
Gene
STAG3
Gene Name
stromal antigen 3
There are no transcripts associated with this gene.

View Full Product Details