Product Details

SNP ID
rs149527213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122077977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAGAAGGATGTCTGATTCCAAA[C/G]CTGTCTCTCATCACAATCATATCTT
Phenotype
MIM: 605113
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AASS PubMed Links

Gene Details

Gene
AASS
Gene Name
aminoadipate-semialdehyde synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005763.3 2668 Missense Mutation AGC,AGG S841R NP_005754.2
XM_011515725.2 2668 Missense Mutation AGC,AGG S809R XP_011514027.1

View Full Product Details