Product Details

SNP ID
rs150043586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100868135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCCTGCAGACAGGGGGGGCCTT[C/T]GCCCTGGAAGCCTGGACGCCGAGAT
Phenotype
MIM: 616861 MIM: 614469 MIM: 602933
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6875 PubMed Links

Gene Details

Gene
MIR6875
Gene Name
microRNA 6875
There are no transcripts associated with this gene.

Gene
SLC12A9
Gene Name
solute carrier family 12 member 9
There are no transcripts associated with this gene.

Gene
SRRT
Gene Name
serrate, RNA effector molecule
There are no transcripts associated with this gene.

Gene
TRIP6
Gene Name
thyroid hormone receptor interactor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003302.2 435 Missense Mutation CGC,TGC R89C NP_003293.2

View Full Product Details