Product Details
- SNP ID
-
rs150333128
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:130381645 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCCCCAGCTGGACTTCTGGCGG[G/T]GGCCTGCCCACCCTGGCTCCCCCAT
- Phenotype
-
MIM: 114850
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CPA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1126899] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CPA1
- Gene Name
- carboxypeptidase A1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001868.3 |
313 |
Missense Mutation |
GGG,TGG |
G55W |
NP_001859.1 |
View Full Product Details