Product Details

SNP ID
rs150622990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44138977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAGCACAGTGACTTGTAGTCGA[C/T]GTTCCCCGCCAGGTCCATGGGTGTC
Phenotype
MIM: 138079 MIM: 613993
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCK PubMed Links

Gene Details

Gene
GCK
Gene Name
glucokinase
There are no transcripts associated with this gene.

Gene
MYL7
Gene Name
myosin light chain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021223.2 565 Missense Mutation ATC,GTC I158V NP_067046.1
XM_005249817.4 565 UTR 3 XP_005249874.1
XM_011515463.2 565 Missense Mutation ATC,GTC I214V XP_011513765.1
XM_011515464.2 565 Missense Mutation ATC,GTC I191V XP_011513766.1
XM_011515465.2 565 UTR 3 XP_011513767.1
XM_017012478.1 565 UTR 3 XP_016867967.1
XM_017012479.1 565 Missense Mutation ATC,GTC I82V XP_016867968.1

View Full Product Details