Product Details

SNP ID
rs150652838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:36154694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGACCTGCTAGCGGAGCAGCAG[C/T]CTCACCACCTGGCCACAGCTGTGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EEPD1 PubMed Links

Gene Details

Gene
EEPD1
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030636.2 1088 Missense Mutation CCT,TCT P124S NP_085139.2
XM_017012657.1 1088 Missense Mutation CCT,TCT P124S XP_016868146.1
XM_017012658.1 1088 Missense Mutation CCT,TCT P124S XP_016868147.1

View Full Product Details