Product Details

SNP ID
rs150672334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:105111848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGGTTCACCTGGATCTGTAAT[A/T]CCTGCTCAAGCACACGGGAAAATAT
Phenotype
MIM: 608444 MIM: 602980
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KMT2E PubMed Links

Gene Details

Gene
KMT2E
Gene Name
lysine methyltransferase 2E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018682.3 4545 Silent Mutation ATA,ATT I1364I NP_061152.3
NM_182931.2 4545 Silent Mutation ATA,ATT I1364I NP_891847.1
XM_005250493.1 4545 Silent Mutation ATA,ATT I1364I XP_005250550.1
XM_011516400.1 4545 Silent Mutation ATA,ATT I1364I XP_011514702.1
XM_017012435.1 4545 Silent Mutation ATA,ATT I1218I XP_016867924.1
Gene
SRPK2
Gene Name
SRSF protein kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278273.1 4545 Intron NP_001265202.1
NM_182691.2 4545 Intron NP_872633.1
NM_182692.2 4545 Intron NP_872634.1
XM_005250549.3 4545 Intron XP_005250606.2
XM_005250550.3 4545 Intron XP_005250607.1
XM_005250551.4 4545 Intron XP_005250608.1
XM_006716098.2 4545 Intron XP_006716161.1
XM_011516536.2 4545 Intron XP_011514838.1
XM_011516538.2 4545 Intron XP_011514840.1
XM_017012560.1 4545 Intron XP_016868049.1
XM_017012561.1 4545 Intron XP_016868050.1
XM_017012562.1 4545 Intron XP_016868051.1
XM_017012563.1 4545 Intron XP_016868052.1
XM_017012564.1 4545 Intron XP_016868053.1
XM_017012565.1 4545 Intron XP_016868054.1
XM_017012566.1 4545 Intron XP_016868055.1

View Full Product Details