Product Details
- SNP ID
-
rs112279855
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:6937244 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTAATATTCTAAAAAGAATGGA[A/G]ACAGTGCTTTCATTCAATAATTTAA
- Phenotype
-
MIM: 601157
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFA4
PubMed Links
Gene Details
- Gene
- DEFA4
- Gene Name
- defensin alpha 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001925.2 |
|
Intron |
|
|
NP_001916.1 |
View Full Product Details