Product Details

SNP ID
rs112739898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:132560922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCAGGAACAGACTTTAACCTCC[C/T]GAGAAGAGTGGTGGCTCAGAGGATA
Phenotype
MIM: 614930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPYR1 PubMed Links

Gene Details

Gene
HPYR1
Gene Name
Helicobacter pylori responsive 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
LRRC6
Gene Name
leucine rich repeat containing 6
There are no transcripts associated with this gene.

View Full Product Details