Product Details

SNP ID
rs112845592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123253327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAATTTTCTTAACTTACATTTC[A/G]GTCATCTGATTTAGACAGCTTCCGT
Phenotype
MIM: 604764
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZHX1 PubMed Links

Gene Details

Gene
ZHX1
Gene Name
zinc fingers and homeoboxes 1
There are no transcripts associated with this gene.

Gene
ZHX1-C8orf76
Gene Name
ZHX1-C8orf76 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204180.1 Intron NP_001191109.1

View Full Product Details