Product Details

SNP ID
rs113899610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86866503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTTTGAGCCATCAAGATGCCGA[C/T]GTCTTCTCTTCCAGCAGCTATTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNBD1 PubMed Links

Gene Details

Gene
CNBD1
Gene Name
cyclic nucleotide binding domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173538.2 56 Missense Mutation ACG,ATG T3M NP_775809.1
XM_017013149.1 56 Missense Mutation ACG,ATG T3M XP_016868638.1

View Full Product Details