Product Details

SNP ID
rs114102667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:48715285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCAAGTAATATCAACAAGATGA[A/T]ACACTAGGAGATCCCAGCTCTTGTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB1 PubMed Links

Gene Details

Gene
EFCAB1
Gene Name
EF-hand calcium binding domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142857.1 1916 UTR 3 NP_001136329.1
NM_024593.3 1916 Intron NP_078869.1
XM_005251303.1 1916 Intron XP_005251360.1
XM_011517589.1 1916 UTR 3 XP_011515891.1
XM_011517590.2 1916 Intron XP_011515892.1
XM_011517591.2 1916 Intron XP_011515893.1
XM_017013826.1 1916 Intron XP_016869315.1
XM_017013827.1 1916 Intron XP_016869316.1
XM_017013828.1 1916 Intron XP_016869317.1

View Full Product Details