Product Details

SNP ID
rs114699150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109967582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCATCAAATATGGGTTTTCTCTT[C/G]TAAAGCTAAGTATTATATTCAATAG
Phenotype
MIM: 608164
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNV1 PubMed Links

Gene Details

Gene
KCNV1
Gene Name
potassium voltage-gated channel modifier subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014379.3 3042 UTR 3 NP_055194.1

View Full Product Details