Product Details
- SNP ID
-
rs114812027
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:85475400 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAAAAAAAAAAAAAAAAAAGCG[A/G]GAGTCAAGAGAGATTTGGGTTGCTG
- Phenotype
-
MIM: 611492
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CA2
PubMed Links
Gene Details
- Gene
- CA2
- Gene Name
- carbonic anhydrase 2
View Full Product Details