Product Details

SNP ID
rs116323544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:57282168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGAGCAGTGATATTTACACTTC[C/T]GCTTAAAGTTAAGTTGCTGATGACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00588 PubMed Links

Gene Details

Gene
LINC00588
Gene Name
long intergenic non-protein coding RNA 588
There are no transcripts associated with this gene.

View Full Product Details