Product Details

SNP ID
rs116707685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:74235367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGGTCACACCCAGGATTTTCAA[A/C]ATATTGTGCTTCTGTAAGAAACATC
Phenotype
MIM: 605266
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
JPH1 PubMed Links

Gene Details

Gene
JPH1
Gene Name
junctophilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317830.1 3464 UTR 3 NP_001304759.1
NM_020647.3 3464 UTR 3 NP_065698.1
XM_005251274.3 3464 UTR 3 XP_005251331.1
XM_005251275.4 3464 UTR 3 XP_005251332.1

View Full Product Details