Product Details

SNP ID
rs116936201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86214085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTTTTAGTCATTCACTTTTATT[A/G]TAGGATCCCCACAAATTCAGTAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC7A13 PubMed Links

Gene Details

Gene
SLC7A13
Gene Name
solute carrier family 7 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138817.2 1817 UTR 3 NP_620172.2
XM_011516867.1 1817 UTR 3 XP_011515169.1

View Full Product Details