Product Details

SNP ID
rs117779254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:120450932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAAAAGTTATGGTCTGTAAGTA[C/T]GGTATATATGCTGTGTCATCTGCTT
Phenotype
MIM: 610200 MIM: 605927
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL13 PubMed Links

Gene Details

Gene
MRPL13
Gene Name
mitochondrial ribosomal protein L13
There are no transcripts associated with this gene.

Gene
MTBP
Gene Name
MDM2 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022045.4 Intron NP_071328.2
XM_011516962.2 Intron XP_011515264.1
XM_011516963.2 Intron XP_011515265.1

View Full Product Details