Product Details

SNP ID
rs118058413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:119883889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTGAGTGATGAAACTATTACA[A/G]CTATAGAGAACAGAACCTTGTCCTT
Phenotype
MIM: 612974
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEPTOR PubMed Links

Gene Details

Gene
DEPTOR
Gene Name
DEP domain containing MTOR-interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283012.1 Intron NP_001269941.1
NM_022783.3 Intron NP_073620.2
XM_017013735.1 Intron XP_016869224.1

View Full Product Details