Product Details

SNP ID
rs137871765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81802443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGAGGACTCCACCTTTAGGATC[C/T]GTTCACCTTCTGTTTCTGACACATC
Phenotype
MIM: 614903
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNX16 PubMed Links

Gene Details

Gene
SNX16
Gene Name
sorting nexin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022133.3 1136 Missense Mutation CAG,CGG Q292R NP_071416.2
NM_152836.2 1136 Missense Mutation CAG,CGG Q292R NP_690049.1
NM_152837.2 1136 Missense Mutation CAG,CGG Q263R NP_690050.1
XM_005251282.4 1136 Missense Mutation CAG,CGG Q263R XP_005251339.1
XM_005251283.2 1136 Missense Mutation CAG,CGG Q263R XP_005251340.1
XM_011517574.2 1136 Missense Mutation CAG,CGG Q135R XP_011515876.1

View Full Product Details