Product Details

SNP ID
rs139422405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143430289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCGGCAGGCGGTGGCAGAAGC[C/G]CTCGGCCTCGGGAGGCTCCTTCTTC
Phenotype
MIM: 610303
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAFA PubMed Links

Gene Details

Gene
MAFA
Gene Name
MAF bZIP transcription factor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201589.3 144 Missense Mutation CGC,GGC R40G NP_963883.2
Gene
ZC3H3
Gene Name
zinc finger CCCH-type containing 3
There are no transcripts associated with this gene.

View Full Product Details