Product Details

SNP ID
rs140712091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144379470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGCTGTGTTCTTCAGGAGAAA[C/T]GTCAGGGGCCTTCCTCCAAGGTAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADCK5 PubMed Links

Gene Details

Gene
ADCK5
Gene Name
aarF domain containing kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174922.4 177 Silent Mutation AAC,AAT N32N NP_777582.4
XM_006716527.2 177 UTR 5 XP_006716590.1
XM_011516907.2 177 Silent Mutation AAC,AAT N106N XP_011515209.1
XM_011516909.1 177 UTR 5 XP_011515211.1
XM_011516910.1 177 UTR 5 XP_011515212.1
XM_017013174.1 177 UTR 5 XP_016868663.1
Gene
LOC101928902
Gene Name
uncharacterized LOC101928902
There are no transcripts associated with this gene.

View Full Product Details