Product Details

SNP ID
rs140874999
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144316677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGCGTTGCCATAGTTGCCCTG[A/G]AAAAAGCGGCCCACGAACCAGGCCA
Phenotype
MIM: 604900 MIM: 140580
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DGAT1 PubMed Links

Gene Details

Gene
DGAT1
Gene Name
diacylglycerol O-acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012079.5 1348 Silent Mutation NP_036211.2
XM_011517356.2 1348 Silent Mutation XP_011515658.1
Gene
HSF1
Gene Name
heat shock transcription factor 1
There are no transcripts associated with this gene.

Gene
MIR6848
Gene Name
microRNA 6848
There are no transcripts associated with this gene.

View Full Product Details