Product Details

SNP ID
rs141021862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23024256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGAGACCTTTCAGCTTCTGCC[A/G]GTTCCTGTAACACATAGTGGGGAAT
Phenotype
MIM: 607352 MIM: 603612
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHOBTB2 PubMed Links

Gene Details

Gene
RHOBTB2
Gene Name
Rho related BTB domain containing 2
There are no transcripts associated with this gene.

Gene
TNFRSF10B
Gene Name
TNF receptor superfamily member 10b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003842.4 1147 Missense Mutation NP_003833.4
NM_147187.2 1147 Missense Mutation NP_671716.2
XM_017013944.1 1147 Missense Mutation XP_016869433.1

View Full Product Details