Product Details

SNP ID
rs141600603
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81459295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATCAACACTAATAGTTACTGTC[A/G]GTTTCACTAACCCTGCCATGTTCCG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FABP9 PubMed Links

Gene Details

Gene
FABP9
Gene Name
fatty acid binding protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080526.1 116 Missense Mutation NP_001073995.1

View Full Product Details