Product Details

SNP ID
rs141736827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144523303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCTGGAGCGCCAGGCGCGGGGG[C/T]TCTGCACACATGATCTTCCTGTCCC
Phenotype
MIM: 615880 MIM: 603780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP39 PubMed Links

Gene Details

Gene
ARHGAP39
Gene Name
Rho GTPase activating protein 39
There are no transcripts associated with this gene.

Gene
C8orf82
Gene Name
chromosome 8 open reading frame 82
There are no transcripts associated with this gene.

Gene
LRRC14
Gene Name
leucine rich repeat containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272036.1 846 UTR 3 NP_001258965.1
NM_014665.3 846 UTR 3 NP_055480.1
XM_005272358.4 846 UTR 3 XP_005272415.1
XM_005272359.4 846 UTR 3 XP_005272416.1
XM_005272360.4 846 UTR 3 XP_005272417.1
XM_017014005.1 846 UTR 3 XP_016869494.1
Gene
LRRC24
Gene Name
leucine rich repeat containing 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024678.3 846 Silent Mutation GAA,GAG E238E NP_001019849.2
Gene
RECQL4
Gene Name
RecQ like helicase 4
There are no transcripts associated with this gene.

View Full Product Details