Product Details
- SNP ID
-
rs142500152
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:138589346 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGGCCTGGGGGACCGGGAGGTCC[C/T]GGGAGGCCAGGATGTCCAGCTGGTC
- Phenotype
-
MIM: 610026
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
COL22A1
PubMed Links
Gene Details
- Gene
- COL22A1
- Gene Name
- collagen type XXII alpha 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152888.2 |
3387 |
Silent Mutation |
CCA,CCG |
P1596P |
NP_690848.1 |
XM_011516883.2 |
3387 |
Silent Mutation |
CCA,CCG |
P1576P |
XP_011515185.1 |
XM_011516884.2 |
3387 |
Silent Mutation |
CCA,CCG |
P1563P |
XP_011515186.1 |
XM_011516885.2 |
3387 |
Silent Mutation |
CCA,CCG |
P1558P |
XP_011515187.1 |
XM_011516886.2 |
3387 |
Silent Mutation |
CCA,CCG |
P1547P |
XP_011515188.1 |
XM_011516887.2 |
3387 |
Silent Mutation |
CCA,CCG |
P1234P |
XP_011515189.1 |
XM_011516888.2 |
3387 |
Intron |
|
|
XP_011515190.1 |
XM_011516889.2 |
3387 |
Silent Mutation |
CCA,CCG |
P1028P |
XP_011515191.1 |
XM_017013150.1 |
3387 |
Silent Mutation |
CCA,CCG |
P1534P |
XP_016868639.1 |
XM_017013151.1 |
3387 |
Silent Mutation |
CCA,CCG |
P1522P |
XP_016868640.1 |
XM_017013152.1 |
3387 |
Silent Mutation |
CCA,CCG |
P1234P |
XP_016868641.1 |
View Full Product Details