Product Details

SNP ID
rs142758819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:89758141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGACCTGCGCTACCTGAGCCGCGG[C/T]GCCTCTGGCACTGTGTCGTCCGCCC
Phenotype
MIM: 603455
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929709 PubMed Links

Gene Details

Gene
LOC101929709
Gene Name
uncharacterized LOC101929709
There are no transcripts associated with this gene.

Gene
RIPK2
Gene Name
receptor interacting serine/threonine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003821.5 359 Silent Mutation GGC,GGT G27G NP_003812.1
XM_005251092.3 359 UTR 5 XP_005251149.1
XM_011517357.2 359 Intron XP_011515659.1

View Full Product Details