Product Details

SNP ID
rs142780506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:90791931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCGCCGTCCTCCAACAACTCCTC[A/G]GAGGAGCTCAGCTCTGCTCTGCACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NECAB1 PubMed Links

Gene Details

Gene
NECAB1
Gene Name
N-terminal EF-hand calcium binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022351.4 164 Silent Mutation TCA,TCG S15S NP_071746.1
XM_011517213.2 164 Silent Mutation TCA,TCG S15S XP_011515515.1

View Full Product Details