Product Details

SNP ID
rs142788403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:20197422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGACAAGATGGCGCTGCGGGCG[A/G]TGCGGGGGATTGTCAACGGGGCCGC
Phenotype
MIM: 606939
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1B2 PubMed Links

Gene Details

Gene
ATP6V1B2
Gene Name
ATPase H+ transporting V1 subunit B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001693.3 230 Missense Mutation ATG,GTG M6V NP_001684.2

View Full Product Details