Product Details

SNP ID
rs143321089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:43056536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAGCCTGGACTCGCCCTCCTAT[A/G]TCCTGTACAGGTAACGCCCCCGCGG
Phenotype
MIM: 134635
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FNTA PubMed Links

Gene Details

Gene
FNTA
Gene Name
farnesyltransferase, CAAX box, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002027.2 Intron NP_002018.1

View Full Product Details