Product Details

SNP ID
rs143648905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:85113966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTGAGTACAAGTTAGCTACAGA[A/G]GCTTCATTTTATTATTCATATGCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRCC1 PubMed Links

Gene Details

Gene
LRRCC1
Gene Name
leucine rich repeat and coiled-coil centrosomal protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033402.4 Intron NP_208325.3
XM_005251318.3 Intron XP_005251375.1
XM_005251319.4 Intron XP_005251376.1
XM_005251320.3 Intron XP_005251377.1
XM_005251322.3 Intron XP_005251379.1
XM_017013920.1 Intron XP_016869409.1
XM_017013921.1 Intron XP_016869410.1
XM_017013922.1 Intron XP_016869411.1
XM_017013923.1 Intron XP_016869412.1
XM_017013924.1 Intron XP_016869413.1
XM_017013925.1 Intron XP_016869414.1
XM_017013926.1 Intron XP_016869415.1
XM_017013927.1 Intron XP_016869416.1
XM_017013928.1 Intron XP_016869417.1

View Full Product Details