Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015243.2 | 5306 | Intron | NP_056058.2 | ||
NM_017890.4 | 5306 | Intron | NP_060360.3 | ||
NM_152564.4 | 5306 | Intron | NP_689777.3 | ||
NM_181661.2 | 5306 | Intron | NP_858047.2 | ||
XM_005250800.3 | 5306 | Intron | XP_005250857.1 | ||
XM_005250801.4 | 5306 | UTR 5 | XP_005250858.1 | ||
XM_006716510.3 | 5306 | Intron | XP_006716573.1 | ||
XM_011516848.2 | 5306 | Intron | XP_011515150.1 | ||
XM_011516849.2 | 5306 | Intron | XP_011515151.1 | ||
XM_011516850.2 | 5306 | Intron | XP_011515152.1 | ||
XM_011516851.2 | 5306 | Intron | XP_011515153.1 | ||
XM_011516852.2 | 5306 | Intron | XP_011515154.1 | ||
XM_011516853.2 | 5306 | Intron | XP_011515155.1 | ||
XM_011516854.2 | 5306 | Intron | XP_011515156.1 | ||
XM_011516859.2 | 5306 | Intron | XP_011515161.1 | ||
XM_011516861.1 | 5306 | Intron | XP_011515163.1 | ||
XM_011516865.1 | 5306 | Intron | XP_011515167.1 | ||
XM_011516866.2 | 5306 | Intron | XP_011515168.1 | ||
XM_017013109.1 | 5306 | Intron | XP_016868598.1 | ||
XM_017013110.1 | 5306 | Intron | XP_016868599.1 | ||
XM_017013111.1 | 5306 | Intron | XP_016868600.1 | ||
XM_017013112.1 | 5306 | Intron | XP_016868601.1 |