Product Details

SNP ID
rs144616063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:90625814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACAATAGCTGCATTCAATTCCA[C/T]TTGAGCTCGATGAACTACATAAAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM64 PubMed Links

Gene Details

Gene
TMEM64
Gene Name
transmembrane protein 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008495.3 1000 Missense Mutation ATG,GTG M334V NP_001008495.2
NM_001146273.1 1000 Missense Mutation ATG,GTG M282V NP_001139745.1

View Full Product Details