Product Details

SNP ID
rs145238683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27461651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCAGATCATTCCAGCTAGGAAC[A/G]GAGGCAGAAAGAGGCCGATGGTCCC
Phenotype
MIM: 118502 MIM: 601212
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRNA2 PubMed Links

Gene Details

Gene
CHRNA2
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000742.3 2041 Missense Mutation NP_000733.2
NM_001282455.1 2041 Missense Mutation NP_001269384.1
XM_005273397.2 2041 Missense Mutation XP_005273454.1
XM_006716282.1 2041 Missense Mutation XP_006716345.1
XM_011544388.1 2041 Missense Mutation XP_011542690.1
XM_011544389.1 2041 Missense Mutation XP_011542691.1
XM_017013003.1 2041 Missense Mutation XP_016868492.1
XM_017013004.1 2041 Missense Mutation XP_016868493.1
Gene
PTK2B
Gene Name
protein tyrosine kinase 2 beta
There are no transcripts associated with this gene.

View Full Product Details