Product Details

SNP ID
rs145365134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109087693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAAGCACATGAGGACCCCCACA[A/G]ACTGCTACCTGGTGAGCCTGGCAGT
Phenotype
MIM: 188545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRHR PubMed Links

Gene Details

Gene
TRHR
Gene Name
thyrotropin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003301.5 270 Missense Mutation AAC,GAC N61D NP_003292.1
XM_011517263.2 270 Missense Mutation AAC,GAC N61D XP_011515565.1

View Full Product Details