Product Details

SNP ID
rs145633647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109087562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAACCAAACACAGCTTCAGCCAC[A/G]AGCAGTGGTGGCCTTAGAATACCAG
Phenotype
MIM: 188545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRHR PubMed Links

Gene Details

Gene
TRHR
Gene Name
thyrotropin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003301.5 139 Missense Mutation CAA,CGA Q17R NP_003292.1
XM_011517263.2 139 Missense Mutation CAA,CGA Q17R XP_011515565.1

View Full Product Details