Product Details
- SNP ID
-
rs145720114
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:37698652 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTATGGACAGAGACTAGCTTCAG[C/T]CTCGGCGCTGGGATACCAGTAACTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF703
PubMed Links
Gene Details
- Gene
- ZNF703
- Gene Name
- zinc finger protein 703
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025069.2 |
1980 |
Missense Mutation |
GCC,GTC |
A584V |
NP_079345.1 |
View Full Product Details