Product Details

SNP ID
rs146122298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143605826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGTGCCACCCGGGGTGCACAC[A/G]TCTGAGCGCAGCTGGGCTGGGTGTT
Phenotype
MIM: 130592 MIM: 616408 MIM: 137020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EEF1D PubMed Links

Gene Details

Gene
EEF1D
Gene Name
eukaryotic translation elongation factor 1 delta
There are no transcripts associated with this gene.

Gene
PYCRL
Gene Name
pyrroline-5-carboxylate reductase-like
There are no transcripts associated with this gene.

Gene
TIGD5
Gene Name
tigger transposable element derived 5
There are no transcripts associated with this gene.

Gene
TSTA3
Gene Name
tissue specific transplantation antigen P35B
There are no transcripts associated with this gene.

View Full Product Details