Product Details

SNP ID
rs146310523
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:48920268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGATGGAGGAGGGGGACTCACTC[G/T]CCCCAAAGATGAGGAGTATCCGGAA
Phenotype
MIM: 602150
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
SNAI2 PubMed Links

Gene Details

Gene
SNAI2
Gene Name
snail family transcriptional repressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003068.4 428 Silent Mutation NP_003059.1

View Full Product Details