Product Details

SNP ID
rs146362624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6936032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGCAGAACGTTAATCGACACG[C/T]GTGCAGCAGTATGTGAAACTCACAC
Phenotype
MIM: 601157 MIM: 600471
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEFA4 PubMed Links

Gene Details

Gene
DEFA4
Gene Name
defensin alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001925.2 407 Silent Mutation NP_001916.1
Gene
DEFA6
Gene Name
defensin alpha 6
There are no transcripts associated with this gene.

View Full Product Details