Product Details

SNP ID
rs147412005
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30791274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGATAACAGTAATTGGGTGCACT[A/G]AAAATGGTAACCACATTCCGATCAT
Phenotype
MIM: 176916
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PPP2CB PubMed Links

Gene Details

Gene
PPP2CB
Gene Name
protein phosphatase 2 catalytic subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009552.1 1195 Silent Mutation TTC,TTT F260F NP_001009552.1

View Full Product Details