Product Details

SNP ID
rs147966564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37844620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAATGTTCTCATCTCCAGTGACA[A/G]TGGAGACAGGGGGTACAGGGCAGAT
Phenotype
MIM: 606823 MIM: 607013
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRA2 PubMed Links

Gene Details

Gene
ADGRA2
Gene Name
adhesion G protein-coupled receptor A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032777.9 1260 Intron NP_116166.9
XM_005273471.3 1260 Intron XP_005273528.1
XM_011544481.2 1260 Intron XP_011542783.1
XM_011544482.2 1260 Intron XP_011542784.1
XM_011544483.2 1260 Intron XP_011542785.1
Gene
BRF2
Gene Name
BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018310.3 1260 Missense Mutation ACT,ATT T377I NP_060780.2

View Full Product Details