Product Details

SNP ID
rs149994926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37762715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAGTCGGGTGCGCATTGCGGG[C/T]GGTGAACGAGCGCGTGCAGCAGGCT
Phenotype
MIM: 611605 MIM: 604436
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERLIN2 PubMed Links

Gene Details

Gene
ERLIN2
Gene Name
ER lipid raft associated 2
There are no transcripts associated with this gene.

Gene
PROSC
Gene Name
proline synthetase cotranscribed homolog (bacterial)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007198.3 963 Missense Mutation GCG,GTG A19V NP_009129.1
XM_017013001.1 963 Missense Mutation GCG,GTG A54V XP_016868490.1

View Full Product Details