Product Details

SNP ID
rs150364969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:104348606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCTAAGTCTTTGGGAGATTTA[C/T]GTGTCTCCAAGAAGCCCCGGATGGA
Phenotype
MIM: 605933
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCSTAMP PubMed Links

Gene Details

Gene
DCSTAMP
Gene Name
dendrocyte expressed seven transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257317.1 133 Silent Mutation TAC,TAT Y18Y NP_001244246.1
NM_030788.3 133 Silent Mutation TAC,TAT Y18Y NP_110415.1
XM_011517321.1 133 Silent Mutation TAC,TAT Y18Y XP_011515623.1
XM_011517324.1 133 Silent Mutation TAC,TAT Y18Y XP_011515626.1
XM_017013878.1 133 Silent Mutation TAC,TAT Y18Y XP_016869367.1
XM_017013879.1 133 Silent Mutation TAC,TAT Y18Y XP_016869368.1
XM_017013880.1 133 Silent Mutation TAC,TAT Y18Y XP_016869369.1

View Full Product Details